The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030754.4 | NP_110381.2 | 4 - 4 | 474 | 98 |
GenBank mRNA | BC020795.1 | - | 4 - 4 | 435 | 98 |
X51440.1 | - | 2 - 2 | 184 | 97 | |
X51445.1 | - | 3 - 3 | 389 | 98 |