Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DOPEY1
Entrez Gene ID
23033
Gene Name
dopey family member 1
Gene Aliases
DOP1, KIAA1117, dJ202D23.2
Chromosome Location
Chr.6: 83067650 - 83171351 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199942.1 NP_001186871.1 25 - 26 5905 120
NM_015018.3 NP_055833.2 25 - 26 5932 120
XM_011535619.2 XP_011533921.1 24 - 25 5897 120
XM_017010559.1 XP_016866048.1 25 - 26 5927 120
XM_017010560.1 XP_016866049.1 24 - 25 5855 120
XM_017010561.1 XP_016866050.1 25 - 26 5879 120
XM_017010562.1 XP_016866051.1 24 - 25 5828 120
XM_017010563.1 XP_016866052.1 25 - 26 5906 120
XM_017010564.1 XP_016866053.1 25 - 26 5906 120
XM_017010565.1 XP_016866054.1 25 - 26 5900 120
XM_017010566.1 XP_016866055.1 25 - 26 5900 120
XM_017010567.1 XP_016866056.1 24 - 25 5828 120
XM_017010568.1 XP_016866057.1 24 - 25 5897 120
XM_017010569.1 XP_016866058.1 23 - 24 5814 120
XM_017010570.1 XP_016866059.1 24 - 25 5828 120
XM_017010572.1 XP_016866061.1 25 - 26 5927 120
XM_017010573.1 XP_016866062.1 25 - 26 5927 120
XM_017010574.1 XP_016866063.1 25 - 26 5927 120
GenBank mRNA AB029040.2 - 8 - 9 2953 120
AK027030.1 - 5 - 6 1699 120
BC144544.1 - 25 - 26 5803 120
BC150619.1 - 25 - 26 5803 120
BX648667.1 - 25 - 26 5930 120

View Full Product Details