Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NLGN2
Entrez Gene ID
57555
Gene Name
neuroligin 2
Gene Aliases
-
Chromosome Location
Chr.17: 7405492 - 7419864 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020795.3 NP_065846.1 1 - 2 527 128

View Full Product Details