Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
XAGE5
Entrez Gene ID
170627
Gene Name
X antigen family member 5
Gene Aliases
CT12.5, GAGED5, XAGE-5
Chromosome Location
Chr.X: 52811295 - 52818301 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130775.2 NP_570131.1 2 - 3 141 128
XM_017029320.1 XP_016884809.1 3 - 4 374 128
GenBank mRNA BC103962.1 - 1 - 2 439 128

View Full Product Details