Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM230
Entrez Gene ID
29058
Gene Name
transmembrane protein 230
Gene Aliases
C20orf30, HSPC274, dJ1116H23.2.1
Chromosome Location
Chr.20: 5068483 - 5113087 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009923.1 NP_001009923.1 4 - 5 473 75
NM_001009924.1 NP_001009924.1 4 - 5 691 75
NM_001009925.1 NP_001009925.1 3 - 4 367 75
NM_014145.4 NP_054864.3 3 - 4 598 75
XM_005260713.3 XP_005260770.1 3 - 4 565 75
XM_011529227.2 XP_011527529.1 3 - 4 588 75
XM_011529229.2 XP_011527531.1 4 - 5 613 75
XM_017027836.1 XP_016883325.1 3 - 4 350 75
GenBank mRNA AK055700.1 - 4 - 5 647 75
AK315606.1 - 3 - 4 357 75
AL357536.1 - 1 - 2 60 75
AY359115.1 - 3 - 4 490 75
BC009768.1 - 3 - 4 351 75
BC009769.1 - 3 - 4 357 75
BC009770.1 - 3 - 4 351 75
BC011990.1 - 3 - 4 338 75
BC015113.1 - 3 - 4 302 75
BC070212.1 - 3 - 4 367 75
BC110408.1 - 3 - 4 329 75

View Full Product Details