Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTIF3
Entrez Gene ID
219402
Gene Name
mitochondrial translational initiation factor 3
Gene Aliases
IF3mt
Chromosome Location
Chr.13: 27435639 - 27450900 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166261.1 NP_001159733.1 3 - 4 584 149
NM_001166262.1 NP_001159734.1 3 - 4 654 149
NM_001166263.1 NP_001159735.1 4 - 5 683 149
NM_152912.4 NP_690876.3 3 - 4 622 149
XM_006719771.3 XP_006719834.1 4 - 5 670 149
XM_006719772.3 XP_006719835.1 3 - 4 901 149
XM_011534957.2 XP_011533259.1 4 - 5 1144 149
XM_011534958.2 XP_011533260.1 4 - 5 928 149
XM_011534959.2 XP_011533261.1 4 - 5 1093 149
XM_011534960.2 XP_011533262.1 4 - 5 758 149
XM_011534961.2 XP_011533263.1 4 - 5 1346 149
XM_011534962.2 XP_011533264.1 4 - 5 923 149
XM_011534963.2 XP_011533265.1 4 - 5 1511 149
XM_017020417.1 XP_016875906.1 3 - 4 528 149
GenBank mRNA AF410851.1 - 2 - 3 489 149
AK094420.1 - 1 - 2 1683 149
BC039599.1 - 3 - 4 575 149
BC046166.1 - 3 - 4 594 149
BM548061.1 - 2 - 3 547 149

View Full Product Details