Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 186195876-186196149 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP4V2
Entrez Gene ID
285440
Gene Name
cytochrome P450 family 4 subfamily V member 2
Gene Aliases
BCD, CYP4AH1
UniGene
Hs.587231
Gene Chromosome Location
Chr.4: 186191520-186213463 on Build GRCh38
Transcripts
NM_207352.3 XM_005262935.3 XM_017008037.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4123809 c.374A>T p.K125M Chr.4: 186196049-186196049 on Build GRCh38
5516324 c.376_377insT p.L127fs*42 Chr.4: 186196051-186196052 on Build GRCh38
6372760 c.372C>A p.Y124* Chr.4: 186196047-186196047 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs755224947 Chr.4: 186196099-186196099 on Build GRCh38
rs374279145 Chr.4: 186195997-186195997 on Build GRCh38
rs139878596 Chr.4: 186195960-186195960 on Build GRCh38
rs368998803 Chr.4: 186196013-186196013 on Build GRCh38
rs933815019 Chr.4: 186195957-186195957 on Build GRCh38
rs866920606 Chr.4: 186196024-186196024 on Build GRCh38
rs1013303492 Chr.4: 186196066-186196066 on Build GRCh38
rs375857768 Chr.4: 186195993-186195993 on Build GRCh38
rs199476188 Chr.4: 186196010-186196010 on Build GRCh38
rs769020128 Chr.4: 186196124-186196124 on Build GRCh38
rs751749088 Chr.4: 186196001-186196001 on Build GRCh38
rs779896273 Chr.4: 186196006-186196007 on Build GRCh38
rs755612419 Chr.4: 186195949-186195949 on Build GRCh38
rs1041538731 Chr.4: 186195959-186195959 on Build GRCh38
rs771753407 Chr.4: 186196044-186196044 on Build GRCh38
rs942467014 Chr.4: 186195919-186195919 on Build GRCh38
rs119103283 Chr.4: 186196007-186196007 on Build GRCh38
rs975174218 Chr.4: 186195929-186195929 on Build GRCh38
rs767908341 Chr.4: 186196102-186196102 on Build GRCh38
rs72646252 Chr.4: 186195916-186195916 on Build GRCh38
rs750267362 Chr.4: 186196098-186196098 on Build GRCh38
rs149684063 Chr.4: 186196042-186196042 on Build GRCh38
rs369170322 Chr.4: 186196117-186196117 on Build GRCh38
rs769884383 Chr.4: 186195943-186195943 on Build GRCh38
rs148330760 Chr.4: 186196028-186196028 on Build GRCh38
rs755065439 Chr.4: 186196006-186196006 on Build GRCh38
rs776440884 Chr.4: 186196081-186196081 on Build GRCh38
rs774007286 Chr.4: 186196037-186196037 on Build GRCh38
rs752819969 Chr.4: 186196112-186196112 on Build GRCh38
rs372662173 Chr.4: 186196040-186196040 on Build GRCh38
rs778488831 Chr.4: 186196023-186196023 on Build GRCh38
rs771216416 Chr.4: 186196034-186196034 on Build GRCh38
rs764016255 Chr.4: 186196079-186196079 on Build GRCh38
rs780492071 Chr.4: 186195945-186195945 on Build GRCh38
rs749785448 Chr.4: 186196003-186196003 on Build GRCh38
rs199476189 Chr.4: 186196075-186196075 on Build GRCh38
rs757068699 Chr.4: 186195962-186195962 on Build GRCh38
rs372944636 Chr.4: 186196119-186196119 on Build GRCh38
rs764970229 Chr.4: 186196088-186196088 on Build GRCh38
rs774964019 Chr.4: 186196056-186196056 on Build GRCh38
rs757602094 Chr.4: 186196125-186196125 on Build GRCh38
rs72646253 Chr.4: 186195966-186195966 on Build GRCh38
rs758103859 Chr.4: 186195987-186195987 on Build GRCh38
rs756675453 Chr.4: 186196018-186196018 on Build GRCh38
rs761616967 Chr.4: 186196083-186196083 on Build GRCh38

View Full Product Details