The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003862.2 | NP_003853.1 | 4 - 5 | 897 | 143 |
GenBank mRNA | AB007422.1 | - | 4 - 5 | 360 | 143 |
AF075292.1 | - | 4 - 5 | 897 | 143 | |
AF211188.1 | - | 4 - 5 | 360 | 143 | |
AY358811.1 | - | 4 - 5 | 904 | 143 | |
BC006245.2 | - | 4 - 5 | 504 | 143 | |
BT019570.1 | - | 4 - 5 | 360 | 143 | |
BT019571.1 | - | 4 - 5 | 360 | 143 |