Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRH2
Entrez Gene ID
5555
Gene Name
proline rich protein HaeIII subfamily 2
Gene Aliases
PRP-1/PRP-2, Pr, pr1/Pr2
Chromosome Location
Chr.12: 10929236 - 10934845 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110213.1 NP_001103683.1 1 - 2 107 81
GenBank mRNA BC095488.1 - 1 - 2 107 81
BC136499.1 - 1 - 2 98 81
BC141916.1 - 1 - 2 106 81
BX538206.1 - 1 - 2 108 81
BX538207.1 - 1 - 2 108 81
BX641094.1 - 1 - 2 111 81
BX647155.1 - 1 - 2 107 81
BX647156.1 - 1 - 2 101 81
BX648436.1 - 1 - 2 108 81
BX648656.1 - 1 - 2 37 81
BX648901.1 - 1 - 2 37 81
CR749583.1 - 1 - 2 105 81
K03202.1 - 1 - 2 116 81

View Full Product Details