The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033121.1 | NP_149112.1 | 4 - 5 | 661 | 98 |
XM_005253980.2 | XP_005254037.1 | 4 - 5 | 701 | 98 | |
XM_005253981.3 | XP_005254038.1 | 4 - 5 | 661 | 98 | |
XM_005253982.3 | XP_005254039.1 | 4 - 5 | 661 | 98 | |
XM_005253984.1 | XP_005254041.1 | 4 - 5 | 457 | 98 | |
XM_011538938.2 | XP_011537240.1 | 4 - 5 | 661 | 98 | |
XM_017020157.1 | XP_016875646.1 | 4 - 5 | 661 | 98 | |
XM_017020158.1 | XP_016875647.1 | 4 - 5 | 661 | 98 | |
XM_017020159.1 | XP_016875648.1 | 4 - 5 | 444 | 98 | |
XM_017020160.1 | XP_016875649.1 | 4 - 5 | 444 | 98 | |
XM_017020161.1 | XP_016875650.1 | 4 - 5 | 444 | 98 | |
XM_017020162.1 | XP_016875651.1 | 4 - 5 | 661 | 98 | |
GenBank mRNA | AF064604.1 | - | 4 - 5 | 567 | 98 |
AK302535.1 | - | 4 - 5 | 502 | 98 | |
AK308923.1 | - | 4 - 5 | 439 | 98 | |
AY364251.1 | - | 4 - 5 | 402 | 98 | |
BC032833.2 | - | 4 - 5 | 502 | 98 |