Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SPATA31A7
- Entrez Gene ID
-
26165
- Gene Name
- SPATA31 subfamily A member 7
- Gene Aliases
-
AEP1, C9orf36A, FAM75A4, FAM75A7, SPATA31A4
- Chromosome Location
-
Chr.9: 61190036 - 61196280 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_015667.2 |
NP_056482.2 |
4 - 4 |
1871 |
109 |
GenBank mRNA |
BC040528.1 |
- |
4 - 4 |
1871 |
109 |
- Gene Symbol
- SPATA31A5
- Entrez Gene ID
-
727905
- Gene Name
- SPATA31 subfamily A member 5
- Gene Aliases
-
FAM75A5
- Chromosome Location
-
Chr.9: 60914372 - 60920653 on Build GRCh38
View Full Product Details