The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159651.1 | NP_001153123.1 | 2 - 3 | 587 | 141 |
GenBank mRNA | BC003114.1 | - | 2 - 3 | 512 | 141 |