The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130715.1 | NP_001124187.1 | 2 - 3 | 242 | 68 |
NM_001130716.1 | NP_001124188.1 | 2 - 3 | 281 | 68 | |
NM_016619.2 | NP_057703.1 | 2 - 3 | 242 | 68 | |
GenBank mRNA | AF208846.1 | - | 2 - 3 | 221 | 68 |
AJ422147.1 | - | 2 - 3 | 223 | 68 | |
AK000140.1 | - | 2 - 3 | 242 | 68 | |
BC012205.1 | - | 2 - 3 | 265 | 68 | |
BI824905.1 | - | 2 - 3 | 286 | 68 | |
DQ007338.1 | - | 2 - 3 | 204 | 68 | |
FJ795024.1 | - | 1 - 2 | 120 | 68 | |
FJ882405.1 | - | 1 - 2 | 120 | 68 |