The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146337.2 | NP_001139809.1 | 3 - 4 | 365 | 74 |
NM_001323062.1 | NP_001309991.1 | 3 - 4 | 365 | 74 | |
NM_001323063.1 | NP_001309992.1 | 3 - 4 | 365 | 74 | |
NM_152406.3 | NP_689619.1 | 3 - 4 | 365 | 74 | |
XM_017009036.1 | XP_016864525.1 | 3 - 4 | 342 | 74 | |
GenBank mRNA | AK074185.1 | - | 3 - 4 | 308 | 74 |
AK095980.1 | - | 3 - 4 | 319 | 74 | |
BC040723.2 | - | 3 - 4 | 332 | 74 | |
BC125093.1 | - | 3 - 4 | 284 | 74 | |
BC125094.1 | - | 3 - 4 | 284 | 74 | |
DA792192.1 | - | 3 - 4 | 365 | 74 |