The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008270.2 | NP_001008271.2 | 3 - 4 | 806 | 72 |
GenBank mRNA | AK172866.1 | - | 3 - 4 | 806 | 72 |
BC137353.1 | - | 3 - 4 | 596 | 72 | |
BC137354.1 | - | 3 - 4 | 596 | 72 | |
BC144604.1 | - | 3 - 4 | 564 | 72 |