The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164257.1 | NP_001157729.1 | 1 - 2 | 74 | 136 |
NM_001191029.1 | NP_001177958.1 | 1 - 2 | 74 | 136 | |
NM_001191031.1 | NP_001177960.1 | 1 - 2 | 74 | 136 | |
XM_005257805.4 | XP_005257862.1 | 1 - 2 | 74 | 136 | |
GenBank mRNA | AK021786.1 | - | 1 - 2 | 74 | 136 |
AK302960.1 | - | 1 - 2 | 136 | ||
DB037384.1 | - | 1 - 2 | 136 |