The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080453.2 | NP_001073922.2 | 7 - 8 | 1056 | 80 |
XM_011515260.1 | XP_011513562.1 | 7 - 8 | 1056 | 80 | |
XM_011515262.2 | XP_011513564.1 | 7 - 8 | 1056 | 80 | |
XM_017011959.1 | XP_016867448.1 | 7 - 8 | 1056 | 80 | |
XM_017011960.1 | XP_016867449.1 | 7 - 8 | 1056 | 80 | |
GenBank mRNA | BM547069.1 | - | 1 - 2 | 66 | 80 |
BM553299.1 | - | 4 - 5 | 485 | 80 |