The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002458.2 | NP_002449.2 | 38 - 39 | 16199 | 77 |
GenBank mRNA | AK303892.1 | - | 5 - 6 | 897 | 77 |
U95031.1 | - | 8 - 9 | 1539 | 77 |