Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
H2BFM
Entrez Gene ID
286436
Gene Name
H2B histone family member M
Gene Aliases
-
Chromosome Location
Chr.X: 104039753 - 104042454 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011530922.2 XP_011529224.1 2 - 3 701 139

View Full Product Details