The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003745.1 | NP_003736.1 | 1 - 2 | 109 | 145 |
GenBank mRNA | AB000734.1 | - | 1 - 2 | 109 | 145 |