The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_194247.2 | NP_919223.1 | 11 - 11 | 3834 | 151 |
XM_005246381.4 | XP_005246438.1 | 11 - 11 | 3740 | 151 | |
XM_006712365.2 | XP_006712428.1 | 10 - 10 | 3693 | 151 |