Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRPV3
Entrez Gene ID
162514
Gene Name
transient receptor potential cation channel subfamily V member 3
Gene Aliases
FNEPPK2, OLMS, VRL3
Chromosome Location
Chr.17: 3510502 - 3557995 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK092557.1 - 2 - 2 2254 93

View Full Product Details