The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042706.2 | NP_001036171.1 | 4 - 4 | 464 | 71 |
GenBank mRNA | DQ309554.1 | - | 4 - 4 | 388 | 71 |