The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_024254.1 | 5 - 5 | 821 | 92 | |
GenBank mRNA | BX282364.1 | - | 3 - 3 | 470 | 92 |