Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLEKHG4B
Entrez Gene ID
153478
Gene Name
pleckstrin homology and RhoGEF domain containing G4B
Gene Aliases
-
Chromosome Location
Chr.5: 54065 - 189972 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_052909.3 NP_443141.3 3 - 4 791 73
XM_006714446.3 XP_006714509.1 5 - 6 1929 73
XM_011513968.2 XP_011512270.1 5 - 6 1929 73
XM_011513969.2 XP_011512271.1 5 - 6 1806 73
XM_011513970.1 XP_011512272.1 5 - 6 1518 73
XM_011513971.2 XP_011512273.1 5 - 6 1874 73
XM_011513972.2 XP_011512274.1 5 - 6 1930 73
XM_011513973.1 XP_011512275.1 5 - 6 1927 73
XM_017009112.1 XP_016864601.1 5 - 6 1930 73
GenBank mRNA AB067496.2 - 3 - 4 791 73
BC008352.1 - 3 - 4 483 73

View Full Product Details