The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004226.3 | NP_004217.1 | 4 - 5 | 798 | 149 |
XM_011512168.2 | XP_011510470.1 | 4 - 5 | 763 | 149 | |
XM_011512169.1 | XP_011510471.1 | 4 - 5 | 796 | 149 | |
XM_011512170.1 | XP_011510472.1 | 5 - 6 | 779 | 149 | |
XM_011512171.2 | XP_011510473.1 | 3 - 4 | 291 | 149 | |
GenBank mRNA | AB011421.1 | - | 4 - 5 | 735 | 149 |
AK312865.1 | - | 4 - 5 | 796 | 149 | |
BC016040.1 | - | 4 - 5 | 737 | 149 | |
CR542210.1 | - | 3 - 4 | 474 | 149 | |
CR542225.1 | - | 3 - 4 | 474 | 149 |