Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MECR
Entrez Gene ID
51102
Gene Name
mitochondrial trans-2-enoyl-CoA reductase
Gene Aliases
CGI-63, FASN2B, NRBF1
Chromosome Location
Chr.1: 29192657 - 29230958 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024732.2 NP_001019903.2 8 - 9 1072 66
NM_016011.3 NP_057095.3 8 - 9 945 66
XM_005245885.2 XP_005245942.1 9 - 10 995 66
XM_005245887.2 XP_005245944.1 10 - 11 1241 66
XM_017001413.1 XP_016856902.1 10 - 11 1016 66
XM_017001414.1 XP_016856903.1 11 - 12 1263 66
XM_017001415.1 XP_016856904.1 10 - 11 1134 66
GenBank mRNA AF151821.1 - 8 - 9 919 66
AK025830.1 - 8 - 9 911 66
AK095099.1 - 8 - 9 1038 66
AK225687.1 - 8 - 9 911 66
AK315274.1 - 8 - 9 932 66
BC001419.2 - 8 - 9 916 66
CR456703.1 - 8 - 9 893 66

View Full Product Details