Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MFSD2A
Entrez Gene ID
84879
Gene Name
major facilitator superfamily domain containing 2A
Gene Aliases
MCPH15, MFSD2, NLS1
Chromosome Location
Chr.1: 39955112 - 39969968 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136493.2 NP_001129965.1 3 - 4 577 101
NM_001287809.1 NP_001274738.1 3 - 4 427 101
NM_032793.4 NP_116182.2 3 - 4 538 101
NR_109896.1 3 - 4 538 101
XM_005271285.1 XP_005271342.1 3 - 4 502 101
XM_011542312.2 XP_011540614.1 3 - 4 505 101
GenBank mRNA AB073383.1 - 3 - 4 502 101
AF289609.1 - 3 - 4 494 101
AK074186.1 - 3 - 4 443 101
AK075183.1 - 3 - 4 505 101
AK093223.1 - 3 - 4 539 101
AK291540.1 - 3 - 4 500 101
AK297991.1 - 3 - 4 391 101
AL834184.1 - 3 - 4 492 101
AY358636.1 - 3 - 4 519 101
BC011587.1 - 3 - 4 512 101
BC092414.1 - 3 - 4 480 101

View Full Product Details