Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATG7
Entrez Gene ID
10533
Gene Name
autophagy related 7
Gene Aliases
APG7-LIKE, APG7L, GSA7
Chromosome Location
Chr.3: 11272324 - 11564704 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136031.2 NP_001129503.2 15 - 16 1921 85
NM_001144912.1 NP_001138384.1 14 - 15 1804 85
NM_006395.2 NP_006386.1 15 - 16 1921 85
XM_006712931.3 XP_006712994.1 15 - 16 1864 85
XM_006712932.3 XP_006712995.1 16 - 17 2128 85
XM_006712933.3 XP_006712996.1 15 - 16 2101 85
XM_011533277.2 XP_011531579.1 17 - 18 2252 85
XM_011533278.2 XP_011531580.1 16 - 17 1957 85
XM_011533279.2 XP_011531581.1 17 - 18 2252 85
XM_011533280.2 XP_011531582.1 17 - 18 2252 85
XM_011533281.2 XP_011531583.1 17 - 18 2252 85
XM_011533282.2 XP_011531584.1 17 - 18 2252 85
XM_011533283.2 XP_011531585.1 17 - 18 2252 85
XM_011533284.2 XP_011531586.1 17 - 18 2252 85
XM_011533285.2 XP_011531587.1 16 - 17 2093 85
XM_011533286.2 XP_011531588.1 17 - 18 2252 85
XM_017005542.1 XP_016861031.1 15 - 16 1838 85
XM_017005543.1 XP_016861032.1 15 - 16 1864 85
XM_017005544.1 XP_016861033.1 16 - 17 2126 85
XM_017005545.1 XP_016861034.1 15 - 16 1906 85
XM_017005546.1 XP_016861035.1 16 - 17 1957 85
XM_017005547.1 XP_016861036.1 17 - 18 2252 85
XM_017005548.1 XP_016861037.1 15 - 16 1864 85
XM_017005549.1 XP_016861038.1 15 - 16 1976 85
XM_017005550.1 XP_016861039.1 14 - 15 1705 85
XM_017005551.1 XP_016861040.1 15 - 16 1864 85
XM_017005552.1 XP_016861041.1 14 - 15 1705 85
XM_017005553.1 XP_016861042.1 12 - 13 1494 85
XM_017005554.1 XP_016861043.1 12 - 13 1494 85
GenBank mRNA AF094516.1 - 14 - 15 1829 85
AK075221.1 - 15 - 16 1848 85
AK298833.1 - 15 - 16 1799 85
AK303694.1 - 14 - 15 1731 85
AK314504.1 - 15 - 16 1838 85
AL122075.1 - 9 - 10 1270 85
BC000091.2 - 15 - 16 1836 85

View Full Product Details