Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATG7
Entrez Gene ID
10533
Gene Name
autophagy related 7
Gene Aliases
APG7-LIKE, APG7L, GSA7
Chromosome Location
Chr.3: 11272324 - 11564704 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136031.2 NP_001129503.2 3 - 4 328 76
NM_001144912.1 NP_001138384.1 3 - 4 328 76
NM_006395.2 NP_006386.1 3 - 4 328 76
XM_006712931.3 XP_006712994.1 3 - 4 271 76
XM_006712932.3 XP_006712995.1 4 - 5 535 76
XM_006712933.3 XP_006712996.1 3 - 4 508 76
XM_011533277.2 XP_011531579.1 5 - 6 659 76
XM_011533278.2 XP_011531580.1 4 - 5 364 76
XM_011533279.2 XP_011531581.1 5 - 6 659 76
XM_011533280.2 XP_011531582.1 5 - 6 659 76
XM_011533281.2 XP_011531583.1 5 - 6 659 76
XM_011533282.2 XP_011531584.1 5 - 6 659 76
XM_011533283.2 XP_011531585.1 5 - 6 659 76
XM_011533284.2 XP_011531586.1 5 - 6 659 76
XM_011533285.2 XP_011531587.1 5 - 6 659 76
XM_011533286.2 XP_011531588.1 5 - 6 659 76
XM_017005542.1 XP_016861031.1 3 - 4 245 76
XM_017005543.1 XP_016861032.1 3 - 4 271 76
XM_017005544.1 XP_016861033.1 4 - 5 533 76
XM_017005545.1 XP_016861034.1 3 - 4 313 76
XM_017005546.1 XP_016861035.1 4 - 5 364 76
XM_017005547.1 XP_016861036.1 5 - 6 659 76
XM_017005548.1 XP_016861037.1 3 - 4 271 76
XM_017005549.1 XP_016861038.1 4 - 5 542 76
XM_017005550.1 XP_016861039.1 3 - 4 271 76
XM_017005551.1 XP_016861040.1 3 - 4 271 76
XM_017005552.1 XP_016861041.1 3 - 4 271 76
GenBank mRNA AF094516.1 - 2 - 3 236 76
AK075221.1 - 3 - 4 255 76
AK298833.1 - 3 - 4 272 76
AK303694.1 - 3 - 4 255 76
AK314504.1 - 3 - 4 245 76
BC000091.2 - 3 - 4 243 76

View Full Product Details