The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253826.1 | NP_001240755.1 | 2 - 3 | 871 | 77 |
NM_001253827.1 | NP_001240756.1 | 2 - 3 | 466 | 77 | |
XM_011533105.1 | XP_011531407.1 | 2 - 3 | 220 | 77 | |
GenBank mRNA | AK022753.1 | - | 2 - 3 | 292 | 77 |
AK122747.1 | - | 2 - 3 | 450 | 77 |