The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253826.1 | NP_001240755.1 | 9 - 10 | 1482 | 124 |
NM_001253827.1 | NP_001240756.1 | 10 - 11 | 1247 | 124 | |
NM_024572.3 | NP_078848.2 | 8 - 9 | 1467 | 124 | |
XM_011533105.1 | XP_011531407.1 | 10 - 11 | 1001 | 124 | |
XM_017004906.1 | XP_016860395.1 | 8 - 9 | 932 | 124 | |
XM_017004907.1 | XP_016860396.1 | 8 - 9 | 943 | 124 | |
XM_017004908.1 | XP_016860397.1 | 8 - 9 | 1226 | 124 | |
GenBank mRNA | AB078144.1 | - | 8 - 9 | 827 | 124 |
AK022753.1 | - | 9 - 10 | 903 | 124 | |
AK074394.1 | - | 2 - 3 | 127 | 124 | |
AK091313.1 | - | 8 - 9 | 837 | 124 | |
AK122747.1 | - | 10 - 11 | 1231 | 124 | |
AK225808.1 | - | 8 - 9 | 1190 | 124 | |
AK298735.1 | - | 9 - 10 | 895 | 124 | |
AY358758.1 | - | 8 - 9 | 1160 | 124 | |
BC010659.1 | - | 8 - 9 | 1467 | 124 | |
Y09324.4 | - | 8 - 9 | 827 | 124 |