Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GMEB1
Entrez Gene ID
10691
Gene Name
glucocorticoid modulatory element binding protein 1
Gene Aliases
P96PIF, PIF96
Chromosome Location
Chr.1: 28668728 - 28715603 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001319674.1 NP_001306603.1 6 - 7 740 82
NM_006582.3 NP_006573.2 6 - 7 770 82
NM_024482.2 NP_077808.1 6 - 7 737 82
XM_011540518.2 XP_011538820.1 6 - 7 873 82
XM_011540519.2 XP_011538821.1 6 - 7 713 82
XM_011540521.2 XP_011538823.1 6 - 7 842 82
XM_017000087.1 XP_016855576.1 6 - 7 888 82
GenBank mRNA AF099013.1 - 5 - 6 634 82
AF173868.1 - 6 - 7 665 82
AF203694.1 - 6 - 7 664 82
AK000892.1 - 6 - 7 736 82
AK023126.1 - 6 - 7 657 82
AK315195.1 - 6 - 7 716 82
AK315436.1 - 6 - 7 674 82
BC001473.2 - 6 - 7 687 82

View Full Product Details