Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ABCB8
- Entrez Gene ID
-
11194
- Gene Name
- ATP binding cassette subfamily B member 8
- Gene Aliases
-
EST328128, M-ABC1, MABC1
- Chromosome Location
-
Chr.7: 151028422 - 151047782 on Build GRCh38
- Gene Symbol
- ASIC3
- Entrez Gene ID
-
9311
- Gene Name
- acid sensing ion channel subunit 3
- Gene Aliases
-
ACCN3, DRASIC, SLNAC1, TNaC1
- Chromosome Location
-
Chr.7: 151048292 - 151052756 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK128129.1 |
- |
4 - 5 |
764 |
68 |
View Full Product Details