The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018010.3 | NP_060480.1 | 9 - 10 | 1293 | 96 |
GenBank mRNA | AB014762.1 | - | 9 - 10 | 1045 | 96 |
AF139576.1 | - | 9 - 10 | 1123 | 96 | |
AF245220.1 | - | 9 - 10 | 1187 | 96 | |
AK001009.1 | - | 9 - 10 | 1132 | 96 | |
AK298854.1 | - | 9 - 10 | 1191 | 96 | |
BC009690.2 | - | 5 - 6 | 460 | 96 | |
BC011899.2 | - | 9 - 10 | 1142 | 96 | |
CR457251.1 | - | 9 - 10 | 1045 | 96 |