The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001293273.1 | NP_001280202.1 | 5 - 6 | 643 | 97 |
NM_020166.4 | NP_064551.3 | 7 - 8 | 902 | 97 | |
NR_120639.1 | 5 - 6 | 669 | 97 | ||
NR_120640.1 | 7 - 8 | 1422 | 97 | ||
XM_006713702.1 | XP_006713765.1 | 6 - 7 | 708 | 97 | |
XM_011512992.2 | XP_011511294.1 | 7 - 8 | 1500 | 97 | |
XM_011512993.1 | XP_011511295.1 | 7 - 8 | 902 | 97 | |
XM_017006874.1 | XP_016862363.1 | 6 - 7 | 1267 | 97 | |
XM_017006875.1 | XP_016862364.1 | 5 - 6 | 504 | 97 | |
GenBank mRNA | AB029826.1 | - | 7 - 8 | 887 | 97 |
AF297332.1 | - | 7 - 8 | 921 | 97 | |
AF310339.1 | - | 7 - 8 | 887 | 97 | |
AF310972.1 | - | 7 - 8 | 888 | 97 | |
AK023051.1 | - | 7 - 8 | 878 | 97 | |
AK294882.1 | - | 5 - 6 | 600 | 97 | |
AK316428.1 | - | 6 - 7 | 653 | 97 | |
AL442091.1 | - | 7 - 8 | 821 | 97 | |
AL556872.3 | - | 7 - 8 | 842 | 97 | |
BC004187.2 | - | 7 - 8 | 887 | 97 | |
BC004214.2 | - | 7 - 8 | 887 | 97 | |
CR749608.1 | - | 5 - 6 | 515 | 97 | |
KU178727.1 | - | 7 - 8 | 755 | 97 |