The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282913.1 | NP_001269842.1 | 7 - 8 | 1039 | 81 |
NM_001282914.1 | NP_001269843.1 | 7 - 8 | 1145 | 81 | |
NM_001282915.1 | NP_001269844.1 | 7 - 8 | 963 | 81 | |
NM_173826.3 | NP_776187.2 | 7 - 8 | 1221 | 81 | |
XM_017006251.1 | XP_016861740.1 | 8 - 9 | 2173 | 81 | |
GenBank mRNA | AK124087.1 | - | 4 - 5 | 460 | 81 |
AK292756.1 | - | 7 - 8 | 1039 | 81 | |
AL832473.1 | - | 7 - 8 | 898 | 81 | |
AL833118.1 | - | 5 - 6 | 635 | 81 | |
BC047231.1 | - | 7 - 8 | 840 | 81 | |
BC101058.2 | - | 8 - 9 | 946 | 81 | |
BC101059.2 | - | 7 - 8 | 822 | 81 | |
BC101060.2 | - | 7 - 8 | 898 | 81 | |
BC101061.2 | - | 7 - 8 | 822 | 81 |