The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135052.3 | NP_001128524.1 | 12 - 13 | 2025 | 84 |
NM_001174167.2 | NP_001167638.1 | 13 - 14 | 2040 | 84 | |
NM_001174168.2 | NP_001167639.1 | 12 - 13 | 1890 | 84 | |
NM_003177.6 | NP_003168.2 | 13 - 14 | 2094 | 84 | |
XM_005252147.3 | XP_005252204.1 | 13 - 14 | 2005 | 84 | |
XM_011518946.2 | XP_011517248.1 | 13 - 14 | 1954 | 84 | |
GenBank mRNA | AK075020.1 | - | 9 - 10 | 1158 | 84 |
AK290927.1 | - | 13 - 14 | 1938 | 84 | |
BC001645.2 | - | 13 - 14 | 1986 | 84 | |
BC002962.1 | - | 13 - 14 | 1986 | 84 | |
BC011399.1 | - | 12 - 13 | 1874 | 84 | |
BX647192.1 | - | 5 - 6 | 4097 | 84 | |
FN391891.1 | - | 12 - 13 | 1872 | 84 | |
FN391892.1 | - | 12 - 13 | 1876 | 84 | |
FN391893.1 | - | 12 - 13 | 1835 | 84 | |
L28824.1 | - | 13 - 14 | 1986 | 84 | |
S80267.1 | - | 12 - 13 | 1840 | 84 | |
Z29630.1 | - | 12 - 13 | 1918 | 84 |