The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320808.1 | NP_001307737.1 | 4 - 5 | 1273 | 119 |
NM_014873.2 | NP_055688.1 | 4 - 5 | 684 | 119 | |
XM_005273362.3 | XP_005273419.1 | 5 - 6 | 832 | 119 | |
XM_011510228.2 | XP_011508530.1 | 5 - 6 | 721 | 119 | |
XM_011510229.2 | XP_011508531.1 | 4 - 5 | 604 | 119 | |
XM_017003004.1 | XP_016858493.1 | 3 - 4 | 1836 | 119 | |
GenBank mRNA | AY561706.1 | - | 3 - 4 | 457 | 119 |
BC034621.1 | - | 4 - 5 | 1274 | 119 | |
D86960.1 | - | 4 - 5 | 684 | 119 |