The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004514.3 | NP_004505.2 | 7 - 8 | 1746 | 69 |
GenBank mRNA | U58196.1 | - | 7 - 8 | 1757 | 69 |
U58197.1 | - | 7 - 8 | 1757 | 69 |