Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COL9A2
Entrez Gene ID
1298
Gene Name
collagen type IX alpha 2
Gene Aliases
DJ39G22.4, EDM2, MED, STL5
Chromosome Location
Chr.1: 40300487 - 40317653 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001852.3 NP_001843.1 9 - 10 540 95
XM_006710365.3 XP_006710428.1 10 - 11 648 95
XM_011540715.2 XP_011539017.1 9 - 10 1116 95
XM_011540716.2 XP_011539018.1 8 - 9 596 95
XM_011540717.2 XP_011539019.1 8 - 9 1456 95
XM_017000332.1 XP_016855821.1 9 - 10 582 95
XM_017000333.1 XP_016855822.1 8 - 9 594 95
GenBank mRNA BC041479.1 - 8 - 9 773 95
BC136326.1 - 9 - 10 533 95
BC136327.1 - 9 - 10 533 95
M95610.1 - 9 - 10 429 95

View Full Product Details