The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144925.2 | NP_001138397.1 | 5 - 6 | 888 | 135 |
NM_002462.4 | NP_002453.2 | 3 - 4 | 447 | 135 | |
XM_005260978.4 | XP_005261035.1 | 3 - 4 | 363 | 135 | |
XM_005260979.2 | XP_005261036.1 | 3 - 4 | 556 | 135 | |
XM_005260980.2 | XP_005261037.1 | 3 - 4 | 309 | 154 | |
XM_005260981.2 | XP_005261038.1 | 2 - 3 | 208 | 135 | |
XM_005260982.2 | XP_005261039.1 | 2 - 3 | 174 | 154 | |
XM_011529568.2 | XP_011527870.1 | 7 - 8 | 897 | 135 | |
XM_017028349.1 | XP_016883838.1 | 5 - 6 | 888 | 154 | |
XM_017028350.1 | XP_016883839.1 | 3 - 4 | 556 | 154 | |
XM_017028351.1 | XP_016883840.1 | 3 - 4 | 390 | 135 | |
XM_017028352.1 | XP_016883841.1 | 3 - 4 | 363 | 135 | |
GenBank mRNA | A00209.1 | - | 2 - 3 | 173 | 135 |
AK096355.1 | - | 5 - 6 | 931 | 135 | |
AK225885.1 | - | 3 - 4 | 306 | 135 | |
BC032602.1 | - | 2 - 3 | 197 | 135 | |
CB151486.1 | - | 3 - 4 | 345 | 135 | |
DA351922.1 | - | 3 - 4 | 438 | 135 | |
M30817.1 | - | 1 - 2 | 116 | 135 | |
M33882.1 | - | 3 - 4 | 251 | 135 |