The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136039.2 | NP_001129511.1 | 6 - 7 | 1044 | 70 |
NM_001142355.1 | NP_001135827.1 | 6 - 7 | 993 | 70 | |
NM_001142356.1 | NP_001135828.1 | 4 - 5 | 840 | 70 | |
NM_021824.3 | NP_068596.2 | 6 - 7 | 1196 | 70 | |
XM_005246742.3 | XP_005246799.1 | 6 - 7 | 1259 | 70 | |
XM_011511582.1 | XP_011509884.1 | 6 - 7 | 1180 | 70 | |
XM_011511583.1 | XP_011509885.1 | 6 - 7 | 1924 | 70 | |
XM_011511584.1 | XP_011509886.1 | 6 - 7 | 1742 | 70 | |
XM_011511585.1 | XP_011509887.1 | 6 - 7 | 1691 | 70 | |
XM_017004630.1 | XP_016860119.1 | 6 - 7 | 1441 | 70 | |
GenBank mRNA | AB038949.1 | - | 6 - 7 | 1157 | 70 |
AF060513.1 | - | 6 - 7 | 953 | 70 | |
AF182416.1 | - | 6 - 7 | 1118 | 70 | |
AF283538.1 | - | 6 - 7 | 999 | 70 | |
AK023378.1 | - | 6 - 7 | 1142 | 70 | |
AK225905.1 | - | 6 - 7 | 2519 | 70 | |
AK314709.1 | - | 6 - 7 | 998 | 70 | |
AL834430.1 | - | 5 - 6 | 819 | 70 | |
AY251943.1 | - | 4 - 5 | 843 | 70 | |
BC007654.2 | - | 5 - 6 | 980 | 70 |