The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004426.1 | NP_001004426.1 | 13 - 14 | 2013 | 80 |
NM_001199562.1 | NP_001186491.1 | 13 - 14 | 1968 | 80 | |
NM_003560.2 | NP_003551.2 | 14 - 15 | 2175 | 80 | |
XM_005261764.2 | XP_005261821.1 | 14 - 15 | 2150 | 80 | |
XM_005261765.1 | XP_005261822.1 | 14 - 15 | 2147 | 80 | |
XM_005261766.1 | XP_005261823.1 | 14 - 15 | 2242 | 80 | |
XM_006724332.3 | XP_006724395.1 | 14 - 15 | 2151 | 80 | |
XM_011530422.1 | XP_011528724.1 | 14 - 15 | 2026 | 80 | |
XM_011530423.1 | XP_011528725.1 | 15 - 16 | 2262 | 80 | |
XM_011530424.1 | XP_011528726.1 | 16 - 17 | 2386 | 80 | |
XM_011530425.1 | XP_011528727.1 | 14 - 15 | 2086 | 80 | |
XM_017028981.1 | XP_016884470.1 | 13 - 14 | 2080 | 80 | |
XM_017028982.1 | XP_016884471.1 | 13 - 14 | 1985 | 80 | |
XM_017028983.1 | XP_016884472.1 | 15 - 16 | 2224 | 80 | |
XM_017028984.1 | XP_016884473.1 | 14 - 15 | 2100 | 80 | |
XM_017028985.1 | XP_016884474.1 | 13 - 14 | 1918 | 80 | |
GenBank mRNA | AF064594.1 | - | 14 - 15 | 2175 | 80 |
AF102988.1 | - | 12 - 13 | 1877 | 80 | |
AF102989.1 | - | 13 - 14 | 2039 | 80 | |
AK001290.1 | - | - | 1466 | 80 | |
AK096521.1 | - | 13 - 14 | 1708 | 80 | |
AK128862.1 | - | 3 - 4 | 2931 | 80 | |
AK290825.1 | - | 13 - 14 | 1970 | 80 | |
AK291212.1 | - | 14 - 15 | 2161 | 80 | |
AL080187.1 | - | 14 - 15 | 2175 | 80 | |
BC036742.1 | - | 14 - 15 | 2169 | 80 | |
BC051904.1 | - | 13 - 14 | 1949 | 80 | |
CR456543.1 | - | 14 - 15 | 2103 | 80 |