Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PLA2G6
- Entrez Gene ID
-
8398
- Gene Name
- phospholipase A2 group VI
- Gene Aliases
-
CaI-PLA2, GVI, INAD1, IPLA2-VIA, NBIA2, NBIA2A, NBIA2B, PARK14, PLA2, PNPLA9, iPLA2, iPLA2beta
- Chromosome Location
-
Chr.22: 38111495 - 38192099 on Build GRCh38
View Full Product Details