The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006810.3 | NP_006801.1 | 15 - 16 | 1504 | 99 |
NR_028444.1 | 14 - 15 | 1348 | 99 | ||
GenBank mRNA | AK054963.1 | - | 4 - 5 | 1452 | 99 |
BC001625.1 | - | 14 - 15 | 1295 | 99 | |
CB158962.1 | - | 2 - 3 | 99 | ||
D49490.1 | - | 15 - 16 | 1404 | 99 |