Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HAAO
Entrez Gene ID
23498
Gene Name
3-hydroxyanthranilate 3,4-dioxygenase
Gene Aliases
3-HAO, HAO
Chromosome Location
Chr.2: 42766198 - 42794359 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012205.2 NP_036337.2 4 - 5 426 87
XM_005264230.3 XP_005264287.1 4 - 5 2174 87
XM_011532730.2 XP_011531032.1 5 - 6 520 87
XM_017003717.1 XP_016859206.1 5 - 6 520 87
GenBank mRNA AK295693.1 - 4 - 5 408 87
BC029510.1 - 4 - 5 409 87
BI602406.1 - 4 - 5 414 87
CR457063.1 - 4 - 5 351 87
DT215851.1 - 4 - 5 428 87
Z29481.1 - 4 - 5 391 87

View Full Product Details