The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077620.2 | NP_001071088.1 | 2 - 3 | 242 | 95 |
NR_033357.1 | 2 - 3 | 313 | 95 | ||
XM_017025013.1 | XP_016880502.1 | 2 - 3 | 2260 | 95 | |
XM_017025014.1 | XP_016880503.1 | 2 - 3 | 2260 | 95 | |
XM_017025015.1 | XP_016880504.1 | 2 - 3 | 2260 | 95 | |
GenBank mRNA | AK125617.1 | - | 2 - 3 | 313 | 95 |
BC043413.1 | - | 2 - 3 | 110 | 95 | |
BC146898.1 | - | 2 - 3 | 183 | 95 | |
BC146904.1 | - | 2 - 3 | 183 | 95 | |
DQ390338.1 | - | 2 - 3 | 249 | 95 |