The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077620.2 | NP_001071088.1 | 3 - 4 | 326 | 130 |
NR_033357.1 | 3 - 4 | 397 | 130 | ||
XM_017025013.1 | XP_016880502.1 | 3 - 4 | 2344 | 130 | |
XM_017025014.1 | XP_016880503.1 | 3 - 4 | 2344 | 130 | |
XM_017025015.1 | XP_016880504.1 | 3 - 4 | 2344 | 130 | |
GenBank mRNA | AK125617.1 | - | 3 - 4 | 397 | 130 |
BC043413.1 | - | 3 - 4 | 194 | 130 | |
BC146898.1 | - | 3 - 4 | 267 | 130 | |
BC146904.1 | - | 3 - 4 | 267 | 130 | |
DQ390338.1 | - | 3 - 4 | 333 | 130 |