Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GOLM1
Entrez Gene ID
51280
Gene Name
golgi membrane protein 1
Gene Aliases
C9orf155, GOLPH2, GP73, HEL46, PSEC0257, bA379P1.3
Chromosome Location
Chr.9: 86026141 - 86100201 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016548.3 NP_057632.2 1 - 2 184 93
GenBank mRNA AF236056.1 - 1 - 2 125 93
AK027717.1 - 1 - 2 147 93
AK075542.1 - 1 - 2 118 93
AY358593.1 - 1 - 2 89 93
BC001740.1 - 1 - 2 99 93

View Full Product Details